Anti-Caspase 4 Rabbit Polyclonal Antibody
77047-740
- Antibody Type:Primary
- Antigen Name:Apoptotic cysteine protease Mih1/TX
- Antigen Symbol:Caspase 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P49662
- Antigen Synonyms:Caspase 4 apoptosis related cysteine peptidase|CASP-4|Caspase4|ICE(rel)II|ICE(rel)-II|Caspase-4 subunit 2|Caspase 4|CASP4|CASP4_HUMAN
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:43 and 22 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Caspase 4 (amino acids 62-111)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77047-740
- Packaging:Plastic vial
Rabbit polyclonal antibody to Caspase 4 (p20,cleaved Gln81) for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000
Type: Primary
Antigen: Caspase 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human