Anti-NF-kB p65 Rabbit Polyclonal Antibody
Catalog # 77047-572
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Avian reticuloendotheliosis viral (v rel) oncogene homolog A
- Antigen Symbol:NF-kB p65
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q04206
- Antigen Synonyms:Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3|Nuclear Factor NF Kappa B p65 Subunit|OTTHUMP00000233473|MGC131774|NF kappa B p65delta3|NFKB3|Nuclear factor of kappa light polypeptide gene enhancer in B cells 3|NF-kB p65|Nuclear factor NF-kappa-B p65 subunit
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:60 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human NF-kappaB p65 (amino acids 247-296)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77047-572
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to NF-kappaB p65 for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:5000
Type: Primary
Antigen: NF-kB p65
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat