Anti-MEK4 Rabbit Polyclonal Antibody
Catalog # 77047-064
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:c Jun N terminal kinase kinase 1
- Antigen Symbol:MEK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P45985
- Antigen Synonyms:Dual specificity mitogen-activated protein kinase kinase 4|JNK activating kinase 1|JNKK1|JNK-activating kinase 1|JNKK|C-JUN N-terminal kinase kinase 1|JNK Activated Kinase 1|MAP kinase kinase 4|Dual specificity mitogen activated protein kinase kinase 4
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:44 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SEK1/MKK4 around the phosphorylation site of Ser80 (amino acids 46-95)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77047-064
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to SEK1 (phospho Ser80) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:5000
Type: Primary
Antigen: MEK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat