Order Entry
United States
ContactUsLinkComponent
Anti-ATF5 Rabbit Polyclonal Antibody
Anti-ATF5 Rabbit Polyclonal Antibody
Catalog # 77046-604
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-ATF5 Rabbit Polyclonal Antibody
Catalog # 77046-604
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A94515-100

Specifications

  • Form:
    Liquid
  • Shipping Temperature:
    Shipped on blue ice at 4 °C
  • Cat. No.:
    77046-604

Specifications

About this item

Rabbit polyclonal antibody to ATF5 (acetyl Lys29) for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000

Type: Primary
Antigen: ATF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat