Anti-PKC alpha + beta + gamma Rabbit Polyclonal Antibody
Catalog # 77046-016
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:AAG6
- Antigen Symbol:PKC alpha + beta + gamma
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P17252
- Antigen Synonyms:Aging associated gene 6|PKC-A|PKC alpha + beta + gamma|PKC beta|KPCA_HUMAN|PKC-alpha|PKCB|PKC gamma|PKCA
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:76 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PKC alpha around the phosphorylation site of Tyr657 (amino acids 623-672)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77046-016
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to PKC alpha (phospho Tyr657) for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:20000
Type: Primary
Antigen: PKC alpha + beta + gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat