Anti-MCSF Receptor Rabbit Polyclonal Antibody
77045-960
- Antibody Type:Primary
- Antigen Name:C FMS
- Antigen Symbol:MCSF Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P07333
- Antigen Synonyms:CD115|CSF 1 R|CSF-1 receptor|CD115 antigen|CFMS|CSF 1R|CD 115|Colony stimulating factor 1 receptor|Colony stimulating factor I receptor
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:107 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human M-CSF Receptor around the phosphorylation site of Tyr723 (amino acids 691-740)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77045-960
- Packaging:Plastic vial
Rabbit polyclonal antibody to M-CSF Receptor (phospho Tyr723) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen: MCSF Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat