Anti-PI 3 Kinase p85 beta Rabbit Polyclonal Antibody
Catalog # 77045-916
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:p85
- Antigen Symbol:PI 3 Kinase p85 beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O00459
- Antigen Synonyms:Phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 2 (p85 beta)|Phosphatidylinositol 3 kinase regulatory subunit polypeptide 2|Phosphatidylinositol 3 kinase regulatory beta subunit|Phosphatidylinositol 3 kinase|P85B_HUMAN|Phosphatidylinositol 3 kinase regulatory subunit beta|Phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta|p85 beta|P85B
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:81 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic phospho-peptide derived from human PI 3-kinase p85beta around the phosphorylation site of Tyr464
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77045-916
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to PI 3-kinase p85beta (phospho Tyr464) for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:5000
Type: Primary
Antigen: PI 3 Kinase p85 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat