Anti-NBPF1 + 9 + 10 + 12 + 14 + 15 + 16 + 20 Rabbit Polyclonal Antibody
Catalog # 77045-072
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q5TAG4, Q6P3W6, Q5SXJ2, Q3BBV0, Q3BBW0, Q3BBV1, Q8N660, Q5TI25
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:31 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human NBPF1/9/10/12/14/15/16/20 (amino acids 115-164)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77045-072
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to NBPF1 + 9 + 10 + 12 + 14 + 15 + 16 + 20 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human