Anti-Phospholipase C beta 3 Rabbit Polyclonal Antibody
77044-374
- Antibody Type:Primary
- Antigen Name:1 phosphatidylinositol 4 5 bisphosphate phosphodiesterase beta 3
- Antigen Symbol:Phospholipase C beta 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q01970
- Antigen Synonyms:Phospholipase C beta 3|Phosphoinositide phospholipase C-beta-3|Phosphoinositide phospholipase C|5-bisphosphate phosphodiesterase beta-3|1-phosphatidylinositol-4|PLC beta 3|Plcb3|Phospholipase C beta 3 (phosphatidylinositol specific)|PLC-beta-3
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:138 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PLCB3 around the phosphorylation site of Ser1105 (amino acids 1071-1120)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77044-374
- Packaging:Plastic vial
Rabbit polyclonal antibody to PLCB3 (phospho Ser1105) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:20000
Type: Primary
Antigen: Phospholipase C beta 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat