Anti-Separase Rabbit Polyclonal Antibody
Catalog # 77043-562
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Caspase like protein ESPL1
- Antigen Symbol:Separase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q14674
- Antigen Synonyms:FLJ46492|Caspase-like protein ESPL1|ESP1|ESP 1|Extra spindle poles like 1|Extra spindle poles-like 1 protein|ESPL 1|ESPL1_HUMAN|Extra spindle poles like 1 protein
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:233 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SEPARASE around the phosphorylation site of Ser801 (amino acids 767-816)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77043-562
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to SEPARASE (phospho Ser801) for WB, IHC, IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IHC, IF, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen: Separase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse