Anti-P70 S6 Kinase beta Rabbit Polyclonal Antibody
77042-890
- Antibody Type:Primary
- Antigen Name:70 kDa ribosomal protein S6 kinase 2
- Antigen Symbol:P70 S6 Kinase beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UBS0
- Antigen Synonyms:p70 ribosomal S6 kinase beta|p70 S6KB|P70 S6 Kinase beta|p70 S6K-beta|p70(S6K) beta|EC 2.7.11.1|p70 S6Kbeta|KS6B2_HUMAN|p70 beta
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:53 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic phospho-peptide derived from human p70 S6 Kinase beta around the phosphorylation site of Ser423
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77042-890
- Packaging:Plastic vial
Rabbit polyclonal antibody to p70 S6 Kinase beta (phospho Ser423) for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:1000
Type: Primary
Antigen: P70 S6 Kinase beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse