Order Entry
United States
ContactUsLinkComponent
 
Anti-Rb Rabbit Polyclonal Antibody
  77042-318
undefined
Anti-Rb Rabbit Polyclonal Antibody
  77042-318
 :  A93444-100

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Exon 17 tumor GOS561 substitution mutation causes premature stop
  • Antigen Symbol:
    Rb
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P06400
  • Antigen Synonyms:
    Osteosarcoma|pRb|PP105|OSRC|P105RB|GOS563 exon 17 substitution mutation causes premature stop|pp110|PPP1R130|p105-Rb
  • Storage Buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
  • Molecular Weight:
    106 kDa
  • Storage Temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping Temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human Retinoblastoma around the phosphorylation site of Ser249 (amino acids 221-270)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
  • Cat. No.:
    77042-318
  • Packaging:
    Plastic vial

 

 

Rabbit polyclonal antibody to Retinoblastoma (phospho Ser249) for IF and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: IF, ELISA
  • Recommended Dilutions: IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:40000

Type: Primary
Antigen: Rb
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat