Anti-Human Lambda Light Chain Mouse Monoclonal Antibody [Clone: 4C2] (PE (Phycoerythrin))
76961-656
- Antibody Type:Primary
- Antigen Symbol:Human Lambda Light Chain
- Clonality:Monoclonal
- Clone:4C2
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 T
- Form:Liquid
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Unfortunately, this information is considered to be commercially sensitive
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76961-656
- Packaging:Glass bottle
Mouse monoclonal [4C2] antibody to Human Lambda Light Chain (PE) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: Human Lambda Light Chain
Clonality: Monoclonal
Clone: 4C2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human