Anti-NKG2D Mouse Monoclonal Antibody [Clone: 1D11] (FITC (Fluorescein Isothiocyanate))
76964-988
- Antibody Type:Primary
- Antigen Name:CD314
- Antigen Symbol:NKG2D
- Clonality:Monoclonal
- Clone:1D11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 T
- Form:Liquid
- Gene ID:UniprotID# P26718
- Antigen Synonyms:KLRK1|NK cell receptor D|Killer cell lectin like receptor subfamily K member 1|KLR|NK lectin-like receptor|Killer cell lectin-like receptor subfamily K member 1|KLRC4 KLRK1 readthrough|CD314 antigen|D12S2489E
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:NKL cell line
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76964-988
- Packaging:Glass bottle
Mouse monoclonal [1D11] antibody to NKG2D (FITC) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: NKG2D
Clonality: Monoclonal
Clone: 1D11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human