Anti-Ikaros Mouse Monoclonal Antibody [Clone: 4E9] (PE (Phycoerythrin))
Catalog # 76956-612
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CLL associated antigen KW 6
- Antigen Symbol:Ikaros
- Clonality:Monoclonal
- Clone:4E9
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Size:100 T
- Form:Liquid
- Gene ID:UniprotID# Q13422
- Antigen Synonyms:hIk 1|Ikaros family zinc finger protein 1|Ikzf1|DNA-binding protein Ikaros|IKAROS family zinc finger 1 (Ikaros)|Ikaros (zinc finger protein)|IK1|IKZF1_HUMAN|Hs.54452
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment corresponding to the C terminus of human Ikaros protein
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76956-612
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [4E9] antibody to Ikaros (PE) for Flow Cytometry with samples derived from Human and Mouse.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: Ikaros
Clonality: Monoclonal
Clone: 4E9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse