Anti-CD42a Mouse Monoclonal Antibody [Clone: GR-P] (FITC (Fluorescein Isothiocyanate))
Catalog # 76959-708
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CD42a
- Antigen Symbol:CD42a
- Clonality:Monoclonal
- Clone:GR-P
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Dog
- Size:100 T
- Form:Liquid
- Gene ID:UniprotID# P14770
- Antigen Synonyms:GP9|Glycoprotein 9|GP IX|GP-IX|GPIX_HUMAN|Glycoprotein IX precursor|Glycoprotein IX (platelet)|Glycoprotein IX|GPIX
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Human acute lymphoblastic leukemia cells
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76959-708
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [GR-P] antibody to CD42a (FITC) for Flow Cytometry with samples derived from Human and Canine.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD42a
Clonality: Monoclonal
Clone: GR-P
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog