Anti-TNF Receptor I Mouse Monoclonal Antibody [Clone: H398]
Catalog # 76968-908
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CD120a
- Antigen Symbol:TNF Receptor I
- Clonality:Monoclonal
- Clone:H398
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P19438
- Antigen Synonyms:TNF R55|p55-R|TBPI|TBP1|p55|FPF|TNF R|p60|MGC19588
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, without Sodium Azide (0.2 µm filter sterilized). Endotoxin level is less than 0.01 EU/µg of the protein, as determined by the LAL test
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full length human CD120a
- Tested Applications:IHC-P, IHC-Fr, Functional studies
- Purification:Protein A chromatography
- Cat. No.:76968-908
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [H398] antibody to TNF Receptor I for Flow Cytometry, IP, IHC-P, IHC-Fr and Functional Studies with samples derived from Human.
- Validated Applications: Flow Cytometry, IP, IHC-P, IHC-Fr, Functional Studies
- Recommended Dilutions: Flow Cytometry: 1-4 µg/ml
Type: Primary
Antigen: TNF Receptor I
Clonality: Monoclonal
Clone: H398
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human