Anti-CD180 Mouse Monoclonal Antibody [Clone: G28-8]
76966-252
- Antibody Type:Primary
- Antigen Name:CD180
- Antigen Symbol:CD180
- Clonality:Monoclonal
- Clone:G28-8
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q99467
- Antigen Synonyms:Ly78|CD180 antigen|MGC126233|lymphocyte antigen-64, radioprotective, 105kDa|lymphocyte antigen 64 radioprotective 105kDa|Lymphocyte antigen 64|F630107B15|LY64|CD180_HUMAN
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Human tonsillar B cells
- Purification:Protein A chromatography
- Cat. No.:76966-252
- Packaging:Plastic vial
Mouse monoclonal [G28-8] antibody to CD180 for Flow Cytometry and IP with samples derived from Human.
- Validated Applications: Flow Cytometry, IP
- Recommended Dilutions: Flow Cytometry: 2 µg/ml
Type: Primary
Antigen: CD180
Clonality: Monoclonal
Clone: G28-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human