Anti-CD162 Mouse Monoclonal Antibody [Clone: TC2] (PE (Phycoerythrin))
Catalog # 76959-556
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CD 162
- Antigen Symbol:CD162
- Clonality:Monoclonal
- Clone:TC2
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 T
- Form:Liquid
- Gene ID:UniprotID# Q14242
- Antigen Synonyms:CD162 antigen|P selectin glycoprotein ligand 1|Cutaneous lymphocyte associated associated antigen|CLA|P-selectin glycoprotein ligand 1|CD162|PSGL-1|P selectin glycoprotein ligand 1 precursor|PSGL 1
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Human thymocytes
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76959-556
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [TC2] antibody to CD162 (PE) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD162
Clonality: Monoclonal
Clone: TC2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human