Anti-Thyroid Stimulating Hormone Mouse Monoclonal Antibody [Clone: TSH-51]
Catalog # 76953-482
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:CGA
- Antigen Symbol:Thyroid Stimulating Hormone
- Clonality:Monoclonal
- Clone:TSH-51
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:LSH alpha|Glycoprotein hormones alpha chain precursor|FSHA|LHA|Thyrotropin alpha chain|HCG|Thyroid Stimulating Hormone|TSH alpha|GPHA1
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Human thyrotropin
- Tested Applications:ICC, RIA
- Purification:Protein A chromatography
- Cat. No.:76953-482
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [TSH-51] antibody to Thyroid Stimulating Hormone for ICC, ELISA and RIA with samples derived from Human.
- Validated Applications: ICC, ELISA, RIA
Type: Primary
Antigen: Thyroid Stimulating Hormone
Clonality: Monoclonal
Clone: TSH-51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human