Anti-IRF2BP2 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to IRF2BP2 for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500-1:1, 000
Type: Primary
Antigen: IRF2BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77208-094
- Antigen Symbol:
- IRF2BP2
- Antigen Name:
- I2BP2_HUMAN
- Antigen Synonyms:
- IRF-2BP2|IRF2 binding protein 2|Interferon regulatory factor 2 binding protein 2|IRF-2-binding protein 2|Interferon regulatory factor 2-binding protein 2|IRF2BP2
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q7Z5L9
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 475-545 of human IRF2BP2 (Q7Z5L9)
- Sequence:
- GGQGAGNTGGLEPVHPASLPDSSLATSAPLCCTLCHERLEDTHFVQCPSVPSHKFCFPCSRQSIKQQGASG
- Form:
- Liquid
- Molecular Weight:
- 61 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together