Anti-BMPR2 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to BMPR2 for WB and ICC/IF with samples derived from Human.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500-1:2, 000, ICC/IF: 1:50-1:200
Type: Primary
Antigen: BMPR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77202-192
- Form:
- Liquid
- Shipping Temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together