Anti-Serum Amyloid A Mouse Monoclonal Antibody [clone: SAA/326]
Catalog # 76966-306
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Amyloid fibril protein AA
- Antigen Symbol:Serum Amyloid A
- Clonality:Monoclonal
- Clone:SAA/326
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P02735, P0DJI8
- Antigen Synonyms:PIG4|Amyloid fibrils|OC|Amyloid protein A|SAA 1|PIG 4|SAA 2|SAA|MGC111216
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:12 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full-length human Serum Amyloid A protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76966-306
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [SAA/326] antibody to Serum Amyloid A for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Serum Amyloid A
Clonality: Monoclonal
Clone: SAA/326
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human