Anti-Intelectin 1 Mouse Monoclonal Antibody [clone: ITLN1/4063]
76967-372
- Antibody Type:Primary
- Antigen Name:Endothelial lectin HL 1
- Antigen Symbol:Intelectin 1
- Clonality:Monoclonal
- Clone:ITLN1/4063
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8WWA0
- Antigen Synonyms:hIntL|Galactofuranose binding lectin|Intelectin 1|FLJ20022|Intelectin|Galactofuranose-binding lectin|Intelectin-1|HL1|Endothelial lectin HL-1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:35 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant fragment, around amino acids 16-141, of human Intelectin 1 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76967-372
- Packaging:Plastic vial
Mouse monoclonal [ITLN1/4063] antibody to Intelectin 1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Intelectin 1
Clonality: Monoclonal
Clone: ITLN1/4063
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human