Anti-MRP1 Mouse Monoclonal Antibody [clone: MRP1/1344]
76954-500
- Antibody Type:Primary
- Antigen Name:ABC 29
- Antigen Symbol:MRP1
- Clonality:Monoclonal
- Clone:MRP1/1344
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P33527
- Antigen Synonyms:Abcc1|ABC29|ATP binding cassette transporter variant ABCC1delta ex13&14|ATP binding cassette subfamily C member 1|ABCC|ATP binding cassette transporter variant ABCC1delta ex13|ATP binding cassette sub family C (CFTR/MRP) member 1|ABCC 1|ATP binding cassette sub-family C member 1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:190 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full-length human ABCC1 protein
- Purification:Protein A/G chromatography
- Cat. No.:76954-500
- Packaging:Plastic vial
Mouse monoclonal [MRP1/1344] antibody to MRP1 for Flow Cytometry and IF with samples derived from Human.
- Validated Applications: Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: MRP1
Clonality: Monoclonal
Clone: MRP1/1344
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human