Anti-Human IgM Heavy Chain Mouse Recombinant Antibody [clone: rIGHM/1623]
Catalog # 76958-484
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:Human IgM Heavy Chain
- Clonality:Recombinant
- Clone:rIGHM/1623
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P01871, P20769
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:50 - 75 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full-length human IGHM protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76958-484
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rIGHM/1623] antibody to Human IgM Heavy Chain for ELISA, Flow Cytometry, IF and IHC-P with samples derived from Human.
- Validated Applications: ELISA, Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Human IgM Heavy Chain
Clonality: Recombinant
Clone: rIGHM/1623
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human