Anti-Apolipoprotein A1 Mouse Monoclonal Antibody [clone: APOA1/3661]
Catalog # 76950-258
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Apo-AI
- Antigen Symbol:Apolipoprotein A1
- Clonality:Monoclonal
- Clone:APOA1/3661
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P02647
- Antigen Synonyms:Apolipoprotein A-I|ApoA-I|APOA1_HUMAN|Apolipoprotein A I|Apolipoprotein AI|Apolipoprotein A1|Apolipoprotein A-I(1-242)|ApoA I|APOA1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:28 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant fragment, around amino acids 83-167, of human APOA1 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76950-258
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [APOA1/3661] antibody to Apolipoprotein A1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Apolipoprotein A1
Clonality: Monoclonal
Clone: APOA1/3661
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human