Anti-Histone H1 Mouse Monoclonal Antibody [clone: 1415-1]
Catalog # 76957-890
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:dJ221C16.2
- Antigen Symbol:Histone H1
- Clonality:Monoclonal
- Clone:1415-1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:3005
- Antigen Synonyms:dJ221C16.5|H1 histone family member N testis specific|H1 histone family member 3|H1 histone family|H1 0 histone|H1 histone family member 0|DmeH1|H1 histone family member 5|H1 histone family member 1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:~30 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Nuclei of human leukemia biopsy cells
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76957-890
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [1415-1] antibody to Histone H1 for Flow Cytometry, IF, WB and IHC-P with samples derived from Human, Mouse and Rat.
- Validated Applications: Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Histone H1
Clonality: Monoclonal
Clone: 1415-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat