Anti-P Cadherin Mouse Monoclonal Antibody [clone: 12H6]
Catalog # 76949-696
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CADH3_HUMAN
- Antigen Symbol:P Cadherin
- Clonality:Monoclonal
- Clone:12H6
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P22223
- Antigen Synonyms:Cadherin-3|Cadherin 3 type 1|Cadherin 3 precursor|Cadherin 3|Calcium dependent adhesion protein placental|CDH3 protein|Cadp|CDH 3|CDH3
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:116 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant human P Cadherin fusion protein
- Purification:Protein A/G chromatography
- Cat. No.:76949-696
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [12H6] antibody to P Cadherin for Flow Cytometry, IF and WB with samples derived from Human.
- Validated Applications: Flow Cytometry, IF, WB
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml
Type: Primary
Antigen: P Cadherin
Clonality: Monoclonal
Clone: 12H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human