Anti-Nidogen Rat Monoclonal Antibody [clone: ELM1]
76958-008
- Antibody Type:Primary
- Antigen Name:Enactin
- Antigen Symbol:Nidogen
- Clonality:Monoclonal
- Clone:ELM1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Guinea pig,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P10493
- Antigen Synonyms:NID 1|NID-1|Nidogen|Entactin|NID|Entactin 1|Nid1|NID1_HUMAN|Ent
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:105 kDa/135 kDa/110 kDa/100 kDa/80 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Murine EHS laminin preparation
- Purification:Protein A/G chromatography
- Cat. No.:76958-008
- Packaging:Plastic vial
Rat monoclonal [ELM1] antibody to Nidogen for ELISA with samples derived from Guinea Pig and Mouse.
- Validated Applications: ELISA
Type: Primary
Antigen: Nidogen
Clonality: Monoclonal
Clone: ELM1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Guinea Pig, Mouse