Anti-CD20 Rabbit Recombinant Antibody [clone: IGEL/1497R]
Catalog # 76952-746
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:APY
- Antigen Symbol:CD20
- Clonality:Recombinant
- Clone:IGEL/1497R
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P11836
- Antigen Synonyms:B-lymphocyte surface antigen B1|B Lymphocyte Cell Surface Antigen B1|Bp35|ATOPY|CD 20|B-lymphocyte antigen CD20|B-lymphocyte cell-surface antigen B1|Bp 35|B lymphocyte antigen CD20
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:33 - 37 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full-length human MS4A1 protein
- Tested Applications:IHC-P
- Purification:Protein A chromatography
- Cat. No.:76952-746
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [IGEL/1497R] antibody to CD20 for Flow Cytometry and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD20
Clonality: Recombinant
Clone: IGEL/1497R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human