Anti-gamma Parvin Mouse Monoclonal Antibody [clone: 8C5.2]
Catalog # 76966-746
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:gamma Parvin
- Clonality:Monoclonal
- Clone:8C5.2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9HBI0
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:37 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:GST-tagged recombinant protein corresponding to human gamma Parvin
- Purification:Protein A/G chromatography
- Cat. No.:76966-746
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [8C5.2] antibody to gamma Parvin for IP and WB with samples derived from Human.
- Validated Applications: IP, WB
- Recommended Dilutions: IP: 1 to 2 µg / 500 µg protein lysate, WB: 1 to 2 µg/ml
Type: Primary
Antigen: gamma Parvin
Clonality: Monoclonal
Clone: 8C5.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human