Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: ITGB3/1713]
Catalog # 76954-660
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:BDPLT16
- Antigen Symbol:Integrin beta 3
- Clonality:Monoclonal
- Clone:ITGB3/1713
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P05106
- Antigen Synonyms:HPA 1|CD61 antigen|GP3A|BDPLT2|CD 61|HPA 4|GPIIIa|GT|CD61
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:105 kDa/90 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant human ITGB3 protein
- Tested Applications:IHC
- Purification:Protein A/G chromatography
- Cat. No.:76954-660
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [ITGB3/1713] antibody to Integrin beta 3 for IHC with samples derived from Human.
- Validated Applications: IHC
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Integrin beta 3
Clonality: Monoclonal
Clone: ITGB3/1713
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human