Anti-beta Amyloid Mouse Monoclonal Antibody [clone: APP/3343]
Catalog # 76954-322
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:A4_HUMAN
- Antigen Symbol:beta Amyloid
- Clonality:Monoclonal
- Clone:APP/3343
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P05067
- Antigen Synonyms:AICD-59|ABETA|AD1|ABPP|AID(50)|AICD-50|AICD-57|AID(57)|AAA
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:4 - 46 kDa (beta - Amyloid)/100 - 125 kDa (Amyloid A4)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant fragment, around amino acids 578-680, of human beta Amyloid protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A chromatography
- Cat. No.:76954-322
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [APP/3343] antibody to beta Amyloid for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: beta Amyloid
Clonality: Monoclonal
Clone: APP/3343
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human