Anti-ERCC1 Mouse Monoclonal Antibody [clone: ERCC1/2318]
76967-878
- Antibody Type:Primary
- Antigen Name:COFS 4
- Antigen Symbol:ERCC1
- Clonality:Monoclonal
- Clone:ERCC1/2318
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P07992
- Antigen Synonyms:Excision repair cross complementing 1|COFS4|DNA excision repair protein ERCC1|DNA excision repair protein ERCC-1|Excision repair protein|ERCC1_HUMAN|Excision repair cross complementation group 1|Excision Repair Cross Complementing Rodent Repair Deficiency Complementation Group 1|DNA excision repair protein ERCC 1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:110 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant fragment, around amino acids 191-281, of human ERCC1 protein. The exact sequence is proprietary.
- Purification:Protein A/G chromatography
- Cat. No.:76967-878
- Packaging:Plastic vial
Mouse monoclonal [ERCC1/2318] antibody to ERCC1 for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, WB
- Recommended Dilutions: WB: 1 to 2 µg/ml
Type: Primary
Antigen: ERCC1
Clonality: Monoclonal
Clone: ERCC1/2318
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human