Anti-HLA F Mouse Monoclonal Antibody [clone: 3D11] (PE (Phycoerythrin))
Catalog # 76953-106
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CDA12
- Antigen Symbol:HLA F
- Clonality:Monoclonal
- Clone:3D11
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P30511
- Antigen Synonyms:HLA F antigen|HLA 5.4|HLA class I molecule|HLA-CDA12|HLAF_HUMAN|HLA class I histocompatibility antigen, alpha chain F|HLA-F|HLA F|HLAF
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze.
- Concentration:0.1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Inclusion body-derived HLA F heavy chain
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76953-106
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [3D11] antibody to HLA F (PE) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: 1 to 5 µg/ml
Type: Primary
Antigen: HLA F
Clonality: Monoclonal
Clone: 3D11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human