Anti-NiR Rabbit Polyclonal Antibody
77077-638
- Antibody Type:Primary
- Antigen Name:NET15
- Antigen Symbol:NiR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Synechocystis sp.
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q55366,P05314
- Antigen Synonyms:Novel INHAT (inhibitor of histone acetyltransferase) repressor|NOC2-like protein|NOC2 like|NET7|NOC2L nucleolar associated transcriptional repressor|NOC2L_HUMAN|NIR|NOC2 like nucleolar associated transcriptional repressor|NOC2L
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol (filter sterilised, without Sodium Azide, and carrier free)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:2 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full length Synechocystis strain 6803 NiR protein, expressed in E. coli
- Purification:Affinity purification
- Cat. No.:77077-638
- Packaging:Plastic vial
Rabbit polyclonal antibody to NiR for WB and ELISA with samples derived from Synechocystis and Spinach.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:1,000 to 1:2,000
Type: Primary
Antigen: NiR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Synechocystis, Spinach