Anti-Cadherin like 26 Rabbit Polyclonal Antibody
Catalog # 77070-546
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CAD26_HUMAN
- Antigen Symbol:Cadherin like 26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8IXH8
- Antigen Synonyms:Cadherin-like protein VR20|Cadherin like 26|RP4-551D2.3|Cdh26|Cadherin-like protein 26|Cadherin 26
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:95 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human CDH26 (amino acids 11-60)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77070-546
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to CDH26 for IF and ELISA with samples derived from Human.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:1000
Type: Primary
Antigen: Cadherin like 26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human