Anti-Apc5 Rabbit Polyclonal Antibody
77067-754
- Antibody Type:Primary
- Antigen Name:2510006G12Rik
- Antigen Symbol:Apc5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UJX4
- Antigen Synonyms:ANAPC5|AA408751|Anaphase promoting complex subunit 5|ANAPC 5|Anaphase-promoting complex subunit 5|Cyclosome subunit 5|AA536819|APC5_HUMAN|AA986414
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:85 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human ANAPC5 (amino acids 151-200)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77067-754
- Packaging:Plastic vial
Rabbit polyclonal antibody to ANAPC5 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Apc5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human