Anti-GPR142 Rabbit Polyclonal Antibody
Catalog # 77070-030
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:G protein coupled receptor 142
- Antigen Symbol:GPR142
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q7Z601
- Antigen Synonyms:Probable G-protein coupled receptor 142|GPR142|GP142_HUMAN|Seven transmembrane helix receptor|G-protein coupled receptor PGR2|G protein coupled receptor PGR2|Probable G protein coupled receptor 142
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:51 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human GPR142 (amino acids 41-90)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77070-030
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to GPR142 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: GPR142
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human