Anti-neurobeachin-like 1 Rabbit Polyclonal Antibody
Catalog # 77075-190
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:A530083I02Rik
- Antigen Symbol:neurobeachin-like 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q6ZS30
- Antigen Synonyms:ALS2CR17|Beach|MGC168834|FLJ26555|Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 17 protein|Amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 16|Amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 17|ALS2CR16|MGC164581
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:150 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human NBEAL1 (amino acids 549-598)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77075-190
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to NBEAL1 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: neurobeachin-like 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human