Anti-Gastric Inhibitory Polypeptide Receptor Rabbit Polyclonal Antibody
Catalog # 77075-634
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:Gastric Inhibitory Polypeptide Receptor
- Antigen Symbol:Gastric Inhibitory Polypeptide Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P48546
- Antigen Synonyms:Gippr|GIPR_HUMAN|GIP R|GIP-R|Gm160|Glucose-dependent insulinotropic polypeptide receptor|Gipr|Glucose dependent insulinotropic polypeptide receptor|Gm1081
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:53 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human GIPR (amino acids 83-132)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77075-634
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to GIPR for WB, IHC and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: Gastric Inhibitory Polypeptide Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human