Anti-OR5K1 Rabbit Polyclonal Antibody
77069-944
- Antibody Type:Primary
- Antigen Name:HSHTPCRX10
- Antigen Symbol:OR5K1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8NHB7
- Antigen Synonyms:Olfactory receptor 5K1|OR5K1|OR5K1_HUMAN|Olfactory receptor family 5 subfamily K member 1|Olfactory receptor OR3-8
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:36 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human OR5K1 (amino acids 56-105)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77069-944
- Packaging:Plastic vial
Rabbit polyclonal antibody to OR5K1 for WB, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:5000
Type: Primary
Antigen: OR5K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human