Anti-OR5W2 Rabbit Polyclonal Antibody
Catalog # 77071-412
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Olfactory receptor 5W2
- Antigen Symbol:OR5W2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8NH69
- Antigen Synonyms:Olfactory receptor, family 5, subfamily W, member 2|OR5W2_HUMAN|OR5W2|OR5W2P|Olfactory receptor, family 5, subfamily W, member 3 pseudogene|Olfactory receptor 5W3|Olfactory receptor OR11-155|OR5W3P
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:35 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human OR5W2 (amino acids 158-207)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77071-412
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to OR5W2 for WB, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: OR5W2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human