Anti-ZAK Rabbit Polyclonal Antibody
Catalog # 77062-306
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:AZK
- Antigen Symbol:ZAK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NYL2
- Antigen Synonyms:Leucine zipper and sterile alpha motif containing kinase|HCCS-4|HCCS 4|Mitogen-activated protein kinase kinase kinase MLT|HCCS4|Human cervical cancer suppressor gene 4 protein|Mitogen activated protein kinase kinase kinase MLT|Leucine zipper- and sterile alpha motif-containing kinase|Cervical cancer suppressor gene 4 protein
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:91 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human ZAK (amino acids 131-180)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77062-306
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ZAK for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: ZAK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse