Anti-CPT1B Rabbit Polyclonal Antibody
77067-776
- Antibody Type:Primary
- Antigen Name:Carnitine O palmitoyltransferase 1B
- Antigen Symbol:CPT1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q92523
- Antigen Synonyms:Carnitine O-palmitoyltransferase 1, muscle isoform|Carnitine palmitoyltransferase 1B (muscle)|Carnitine O palmitoyltransferase I muscle isoform|Carnitine O palmitoyltransferase I mitochondrial muscle isoform|Carnitine O-palmitoyltransferase I|Carnitine palmitoyltransferase 1B|Carnitine palmitoyltransferase I muscle|Carnitine palmitoyltransferase I like protein|Carnitine palmitoyltransferase 1A (muscle)
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:87 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human CPT1B (amino acids 387-436)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77067-776
- Packaging:Plastic vial
Rabbit polyclonal antibody to CPT1B for WB, IHC and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: CPT1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human