Anti-DHX8 Rabbit Polyclonal Antibody
Catalog # 77067-808
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ATP dependent RNA helicase DHX8
- Antigen Symbol:DHX8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q14562
- Antigen Synonyms:DEAD/H (Asp Glu Ala Asp/His) box polypeptide 8|DEAH (Asp Glu Ala His) box polypeptide 8|DEAD/H box polypeptide 8|ATP-dependent RNA helicase dhx8|DDX 8|DEAD/H box 8|DDX8|DEAH box polypeptide 8|DEAD/H (Asp Glu Ala Asp/His) box polypeptide 8 (RNA helicase)
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:139 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human DHX8 (amino acids 391-440)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77067-808
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to DHX8 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: DHX8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse