Order Entry
United States
ContactUsLinkComponent
 
Anti-EPS8 like protein 3 Rabbit Polyclonal Antibody
Catalog # 77074-500
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-EPS8 like protein 3 Rabbit Polyclonal Antibody
Catalog # 77074-500
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A99300-100

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Epidermal growth factor receptor kinase substrate 8 like protein
  • Antigen Symbol:
    EPS8 like protein 3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8TE67
  • Antigen Synonyms:
    Epidermal growth factor receptor kinase substrate 8 like protein 3|EPS8L3|Epidermal growth factor receptor pathway substrate 8 related protein 3|EPS8 related protein 3|EPS8R3|EPS8 like 3|EPS8 like protein 3
  • Storage Buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
  • Molecular Weight:
    66 kDa
  • Storage Temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping Temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human EPS8L3 (amino acids 401-450)
  • Tested Applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Cat. No.:
    77074-500
  • Packaging:
    Plastic vial

Specifications

About this item

Rabbit polyclonal antibody to EPS8L3 for IHC, IF and ELISA with samples derived from Human.

  • Validated Applications: IHC, IF, ELISA
  • Recommended Dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:10000

Type: Primary
Antigen: EPS8 like protein 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human