Anti-PSGR Rabbit Polyclonal Antibody
77062-268
- Antibody Type:Primary
- Antigen Name:HPRAJ
- Antigen Symbol:PSGR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9H255
- Antigen Synonyms:Olr59|OR51E3P|Olfactory receptor family 51 subfamily E member 2|Or51e2|Olfactory receptor 51E2|O51E2_HUMAN|Olfactory receptor family 51 subfamily E member 3 pseudogene|Olfactory receptor OR11-16|OR52A2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:35 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human OR51E2 (amino acids 188-237)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77062-268
- Packaging:Plastic vial
Rabbit polyclonal antibody to OR51E2 for WB and ELISA with samples derived from Human and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: PSGR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat