Anti-cadherin 10 Rabbit Polyclonal Antibody
77064-888
- Antibody Type:Primary
- Antigen Name:CAD10_HUMAN
- Antigen Symbol:cadherin 10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9Y6N8
- Antigen Synonyms:T2-cadherin|Cadherin 10 (T2 cadherin)|CDH10|Cadherin 10 type 2|Cadherin10|Cadherin 10 type 2 (T2 cadherin)|cadherin 10|Cadherin-10|T2 cadherin
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:88 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human CDH10 (amino acids 201-250)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77064-888
- Packaging:Plastic vial
Rabbit polyclonal antibody to CDH10 for IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:40000
Type: Primary
Antigen: cadherin 10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse